Learn More
Abcam DAF-FM DA, Nitric o x ide quantification reagent, 5MG
Supplier:  Abcam AB1452955MG
Encompass_Preferred
MW 496.42 g/mol, Purity >98%. Cell permeable reagent for quantification of low concentrations of nitric oxide in solution. Essentially nonfluorescent until it is hydrolyzed to DAF-FM (ab145297) by intracellular esterases and reacts with NO to form a fluorescent benzotriazole.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-258-3259