Learn More
Indofine Chemical Daidzin, 96%, 5 Mg
SDP
Supplier:  Indofine Chemical 021096 5 MG
Encompass_Preferred
DAIDZIN 5 MG, Isoflavone, Daidzein-7-O-glucoside, 7-Glucocosyl-4'-hydroxyisoflavone, C21H20O9, MW: 416.37, 96+%, 552-66-9, MFCD00017466
Catalog No. 50-908-543