Learn More
Selleck Chemical LLC Dalasetron Mesylate Hydrate E4903-100MG
SDP
Supplier:  Selleck Chemical LLC E4903100MG
Encompass_Preferred
Dolasetron Mesylate hydrate(MDL-73147EF hydrate) is an antagonist of serotonin 5-HT3 receptor It is used in the treatment of patients with nausea and vomiting following chemotherapy
Catalog No. 50-000-41731