Learn More
Medchemexpress LLC Dalbergin | 482-83-7 | 25 MG

Supplier: Medchemexpress LLC 25MGHYN8347
Dalbergin is a compound isolated from the knot wood extract of *Dalbergia Sissoo Linn.* It has demonstrated significant antioxidant capabilities and is intended for research use only.
- Isolated from *Dalbergia Sissoo Linn.* knot wood.
- Demonstrates notable antioxidant ability.
- Purity: 98.74%.
- Appearance: Light yellow to yellow solid.
- Molecular formula: C16H12O4.
- Molecular weight: 268.26.
- Solubility: Soluble in DMSO at 100 mg/mL.
- Storage (solid): 4°C, sealed, away from moisture and light.
- Storage (in solvent): -80°C for 6 months, -20°C for 1 month.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.