Learn More
Cayman Chemical DalfoprIstInmesylate 5mg
SDP
Supplier:  Cayman Chemical 197625 MG
Encompass_Preferred
A streptogramin antibiotic that is usually combined with another streptogramin antibiotic, quinupristin, to synergistically kill Gram-positive bacteria
Catalog No. 50-298-2534