Learn More
Medchemexpress LLC Dalmelitinib | 1637658-98-0 | 98.7% | 461.47 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY147259
Encompass_Preferred
Dalmelitinib is an orally active, selective c-Met kinase inhibitor that binds to the ATP-binding region of c-Met. It induces MET phosphorylation and inhibits AKT and ERK phosphorylation. Dalmelitinib potently inhibits cancer cell proliferation and is used in research for cancers such as human non-small cell lung cancer (NSCLC).
  • Orally active selective c-Met kinase inhibitor
  • Binds to the ATP-binding region of c-Met
  • Induces phosphorylation of MET
  • Inhibits phosphorylation of AKT and ERK
  • Potently inhibits cancer cell proliferation
  • Used for research of human non-small cell lung cancer
Catalog No. 50-003-63667