Learn More
Medchemexpress LLC Dalmelitinib | 1637658-98-0 | 98.7% | 461.47 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1472595MG
Encompass_Preferred
Dalmelitinib is an orally active, selective c-Met kinase inhibitor. It induces the phosphorylation of MET and partially or completely inhibits the phosphorylation of AKT and ERK. This compound potently inhibits the proliferation of cancer cells with c-Met oncogene amplification and is used for research in cancers such as human non-small cell lung cancer (NSCLC).
  • Orally active selective c-Met kinase inhibitor (IC50: 2.9 nM)
  • Binds to the ATP-binding region of c-Met
  • Inhibits cancer cell proliferation
  • Used for research in human non-small cell lung cancer
Catalog No. 50-003-63758