Learn More
Medchemexpress LLC Dalmelitinib | 1637658-98-0 | 98.7% | 461.47 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY14725950MG
Encompass_Preferred
Dalmelitinib is an orally active selective c-Met kinase inhibitor that binds to the ATP-binding region of c-Met. It potently inhibits the proliferation of cancer cells with c-Met oncogene amplification and is used for research in cancers such as human non-small cell lung cancer (NSCLC).
  • Orally active selective c-Met kinase inhibitor
  • Binds to the ATP-binding region of c-Met
  • Induces phosphorylation of MET
  • Partially or completely inhibits phosphorylation of AKT and ERK
  • Potently inhibits cancer cell proliferation (c-Met oncogene amplification)
  • Used for research of human non-small cell lung cancer (NSCLC)
Catalog No. 50-003-62814