Learn More
Medchemexpress LLC Dalpiciclib hydrochl 10mM 1mL | 2891598-76-6 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY114338A10MM 1ML
Encompass_Preferred
Dalpiciclib hydrochloride is an orally active, selective inhibitor of cyclin-dependent kinases 4 and 6 (CDK4/6) used in research to study cell-cycle regulation and anticancer activity. It is provided as a 10 mM solution in DMSO and as solid quantities for in vitro experiments. Reported IC50 values are 12.4 nM (CDK4) and 9.9 nM (CDK6).
  • Orally active, selective CDK4/6 inhibitor with low nanomolar potency.
  • Available as 10 mM solution in DMSO and as various solid quantities for flexibility.
  • High reported purity (98.2%) suitable for research applications.
  • Molecular formula C25H31ClN6O2 and molecular weight 483.01.
  • Demonstrated antitumor activity in breast and esophageal cancer models.
Catalog No. 50-002-29163