Learn More
Medchemexpress LLC Dalutrafusp alfa 5mg | 5mg
SDP
Supplier:  Medchemexpress LLC HYP998415MG
Encompass_Preferred
Dalutrafusp alfa (AGEN-1423 GS-1423) is a specific bifunctional antibody against CD73 and TGF- which is involved in the immunosuppressive pathway[1]
Catalog No. 50-003-21733