Learn More
AdipoGen DAMGO . acetate (1 mg)
SDP
Supplier:  AdipoGen AGCP30005VM001
Encompass_Preferred
DAMGO . acetate (1 mg), AdipoGen Life Sciences. Chemical. CAS: 100929-53-1. Formula: C26H35N5O6 . C2H4O2. MW: 513.6 . 60.1. Synthetic. Potent, selective agonist of µ-opioid receptor. Antinociceptive. Stimulates calcium-activated adenylyl cyclases related cAMP production. Expresses chemokines and chemokine receptors through TGF-beta1. Increases food intake of rats.
Catalog No. 50-149-0027