Learn More
Cayman Chemical DamInozIde-d4 5mg
SDP
Supplier:  Cayman Chemical 392645 MG
Encompass_Preferred
An internal standard for the quantification of daminozide by GC- or LC-MS
Catalog No. 50-291-9103