Learn More
Medchemexpress LLC Daminozide (Standard) | 1596-84-5 | MFCD00002787 | 99.7% | 160.17 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY13643R100MG
Encompass_Preferred
Daminozide (Standard) is the analytical standard of Daminozide, intended for research and analytical applications. It functions as a plant growth regulator and a selective inhibitor of human KDM2/7 histone demethylases. It demonstrates significant selectivity for the KDM2/7 subfamily over other tested demethylase subfamily members.
  • Analytical standard for research and analytical applications
  • Selective inhibitor of human KDM2/7 histone demethylases
  • Used in qualitative, quantitative, and methodological research experiments such as HPLC, GC, and MS
Catalog No. 50-003-50370