Learn More
Medchemexpress LLC Danicamtiv | 1970972-74-7 | 99.6% | 435.42 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY10912810MG
Encompass_Preferred
Danicamtiv is an inotropic agent and a selective allosteric activator of cardiac myosin. It increases cardiac systolic function and preserves mechanical efficiency. This product is for research use only.
  • Selective allosteric activator of cardiac myosin.
  • Increases cardiac systolic function.
  • Preserves mechanical efficiency.
  • Selectively enhances cardiac actomyosin activity, prolonging contraction while preserving actin-myosin detachment, allowing relaxation, and without impacting Ca2+ homeostasis.
Catalog No. 50-003-45500