Learn More
Medchemexpress LLC Danicamtiv | 1970972-74-7 | 99.6% | 435.42 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY109128100MG
Encompass_Preferred
Danicamtiv is an inotropic agent that acts as a selective allosteric activator of cardiac myosin. It works by increasing cardiac systolic function and preserving mechanical efficiency.
  • Increases cardiac systolic function.
  • Preserves mechanical efficiency.
  • Selectively activates cardiac myosin.
  • Enhances cardiac actomyosin activity.
  • Prolongs cardiac contraction while allowing relaxation.
  • Does not impact calcium homeostasis.
Catalog No. 50-003-43312