Learn More
Selleck Chemical LLC Danicopan S0803-10MM/1ML
SDP
Supplier:  Selleck Chemical LLC S080310MM/1ML
Encompass_Preferred
Danicopan is a potent selective and orally active inhibitor of factor D with Kd of 0 54 nM for human Factor D Danicopan inhibits alternative pathway of complement (APC) activity and has potential to block the alternative pathway of complement in paroxysmal nocturnal hemoglobinuria (PNH) and atypical hemolytic uremic syndrome (aHUS)
Catalog No. 50-000-41818