Learn More
Medchemexpress LLC Danofloxacin mesylate | 119478-55-6 | 99.9% | 1 ML
SDP
Supplier:  Medchemexpress LLC HYB050110MM/1ML
Encompass_Preferred
Danofloxacin mesylate is a potent fluoroquinolone antibiotic primarily utilized as a veterinary drug. It exhibits broad-spectrum bactericidal activity by targeting bacterial DNA gyrase, an essential enzyme for bacterial replication. Furthermore, this compound functions as a substrate for ATP-dependent efflux transporters, including P-glycoprotein (P-gp) and Multidrug Resistance-associated Protein 2 (MRP2), which are involved in drug transport mechanisms within cells.
  • Fluoroquinolone antibiotic for veterinary use
  • Possesses broad-spectrum bactericidal activity
  • Inhibits bacterial DNA gyrase
  • Acts as a substrate for ATP-dependent efflux transporters
Catalog No. 50-004-06839