Learn More
Medchemexpress LLC HY-10238 10mg Medchemexpress, Danoprevir CAS:850876-88-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1023810MG
Encompass_Preferred
Medchemexpress, HY-10238 10mg Danoprevir CAS:850876-88-9 Danoprevir (ITMN-191) is an orally active NS3/4A protease inhibitor for hepatitis C virus (HCV) with an IC50 of 0.29 nM and is selective for NS3/4A over a panel of 53 proteases (IC50 higher than 10 μM). Danoprevir (ITMN-191) inhibits HCV genotypes 1a, 1b, 4, 5, and 6 (IC50s=0.2-0.4 nM) as well as 2b and 3a (IC50s=1.6, 3.5 nM)[2]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8709