Learn More
Medchemexpress LLC Dansyl chloride | 605-65-2 | 100.0% | 10 G
SDP
Supplier:  Medchemexpress LLC HYD001710G
Encompass_Preferred
Dansyl chloride is a reagent that produces stable blue or blue-green fluorescent sulfonamide adducts in the reaction of aliphatic and aromatic amines with primary amino groups, and is widely used for modified amino acids, protein sequencing and amino acid analysis.
  • Provides fluorescent labeling for protein studies.
  • Reacts with α-chymotrypsin for studying α-amino and other reactive groups in proteins and peptides.
  • Useful for determining the amino acid sequences of small peptides.
  • Reacts with various bases to form derivatives.
Catalog No. 50-004-10427