Learn More
Medchemexpress LLC Dansyl-L-phenylalanine | 1104-36-5 | MFCD00037730 | >98.0% | 398.48 g/mol | C21H22N2O4S | 5 G

Supplier: Medchemexpress LLC HY1183495G
Dansyl-L-phenylalanine is a dansylated derivative of L-phenylalanine that contains a 5-dimethylaminonaphthalene-1-sulfonyl (dansyl) fluorophore attached to the amino group. It is used as a fluorescent probe or label in biochemical and analytical applications and is supplied as a solid.
- Fluorescent dansyl label for detection and spectroscopy
- Used in biochemical assays and probe synthesis
- CAS 1104-36-5; chemical formula C21H22N2O4S
- Molecular weight ~398.48 g/mol
- Typical purity >98.0%
- Common pack sizes: 1 G, 5 G, 10 G
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.