Learn More
U.S. Pharmacopeia DANTROLENE RELATED COMPOUND C
Supplier:  U.S. Pharmacopeia 1163184
Encompass
Dantrolene Related Compound C (50 mg) (5-(4-Nitrophenyl)furan-2-carbaldehyde)Current Lot: F0G099CAS RN® 7147-77-5Harmonized System (HS) Code *: 293219UN No: N/ANDC No: N/AMolecular Formula: C11H7NO4Container Type: VIALBase Control Substance (substance %): N/A
Catalog No. NC2185991