Learn More
Medchemexpress LLC Dantrolene sodium | 14663-23-1 | 99.8% | 336.23 | 500 MG
SDP
Supplier:  Medchemexpress LLC HY14657500MG
Encompass_Preferred
Dantrolene sodium is an orally active, non-competitive glutathione reductase and calcium channel protein inhibitor. It inhibits Ca2+ release from RyR1 and RyR3, benefiting pathologies involving calcium homeostasis disruption, such as stroke and neurodegenerative diseases. It also relieves muscle spasms and malignant hyperthermia, with antitoxic, antipyretic, and anti-inflammatory properties.
  • Orally active glutathione reductase and calcium channel protein inhibitor
  • Inhibits Ca2+ release from RyR1 and RyR3
  • Beneficial for calcium homeostasis disruptions
  • Relieves muscle spasms and malignant hyperthermia
  • Possesses antitoxic, antipyretic, and anti-inflammatory properties
Catalog No. 50-003-63007