Learn More
Medchemexpress LLC DAO-IN-2 250mg | 39793-31-2 | 250 MG
SDP
Supplier:  Medchemexpress LLC HY117292250MG
Encompass_Preferred
DAO-IN-2 (4H-thieno[3,2-b]pyrrole-5-carboxylic acid) is a small-molecule research reagent that inhibits d-amino acid oxidase (DAO) and is used in biochemical and pharmacological studies. It is supplied as a high-purity solid appropriate for in vitro and in vivo experiments exploring DAO-related mechanisms.
  • Moderate potency against human DAO (reported h-DAO IC50 ≈ 245 nM)
  • High reported purity suitable for analytical work and biological assays
  • Supplied as a stable solid for reliable dosing and storage
  • Available in multiple pack sizes to support screening and scale-up studies
  • Intended for research use only; not for human consumption
Catalog No. 50-002-28539