Learn More
Medchemexpress LLC Dapaconazole | 1269726-67-1 | 99.8% | 415.24 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY1671950MG
Encompass_Preferred
Dapaconazole is an antifungal agent known for its ability to inhibit sterol 14α-demethylase cytochrome P450 activity. This compound has demonstrated efficacy in inhibiting key fungal enzymes and has been investigated in clinical trials for conditions such as Tinea Cruris and Tinea Pedis. It is characterized by high purity and specific chemical properties.
  • Inhibits sterol 14α-demethylase cytochrome P450 activity
  • Investigated in clinical trials for antifungal applications
  • High purity
  • Solid appearance
Catalog No. 50-003-93591