Learn More
Medchemexpress LLC Dapagliflozin ((2S)-1,2-propanediol, hydrate) | 960404-48-2 | 100.0% | 502.98 | C24H35ClO9 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY10450A100MG
Encompass_Preferred
Dapagliflozin ((2S)-1,2-propanediol, hydrate) is the propanediol monohydrate form of a selective sodium-glucose cotransporter 2 (SGLT2) inhibitor supplied as a solid for research and analytical use in diabetes and metabolic studies. The material is analytically characterized and provided in laboratory-scale pack sizes for experimental workflows.
  • Selective SGLT2 inhibitor for metabolic and diabetes research.
  • High purity by HPLC (99.99%).
  • Appearance: white to off-white solid.
  • Supplied in convenient laboratory pack sizes including 100 MG.
  • Analytical identity confirmed by 1H NMR and LCMS.
  • Store sealed, away from moisture; in solvent store at -80°C (up to 2 years) or -20°C (up to 1 year).
Catalog No. 50-002-41747