Learn More
Medchemexpress LLC Dapagliflozin ((2S)-1,2-propanediol, hydrate) | 960404-48-2 | MFCD28167768 | 100.0% | 502.98 g·mol⁻¹ | C24H35ClO9 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY10450A10MG
Encompass_Preferred
Dapagliflozin ((2S)-1,2-propanediol, hydrate) is the propanediol monohydrate form of dapagliflozin, a selective sodium-glucose cotransporter 2 (SGLT2) inhibitor provided for research use. It is intended for in vitro and preclinical studies of glucose transport, metabolic pathways, and pharmacology; consult the certificate of analysis and safety data sheet for handling, storage, and purity details.
  • Propanediol monohydrate form for consistent solubility and handling.
  • Selective SGLT2 inhibitor for studies of glucose transport and metabolism.
  • High purity (99.99% HPLC) suitable for analytical and preclinical applications.
  • Molecular weight 502.98 g·mol⁻¹ and formula C24H35ClO9 for accurate dosing and calculations.
  • Available as small solid quantities and as concentrated solution stocks for flexible dosing.
Catalog No. 50-002-43093