Learn More
Medchemexpress LLC Dapagliflozin ((2S)-1,2-propanediol, hydrate) | 960404-48-2 | MFCD28167768 | 100.0% | 502.98 g·mol⁻¹ | C24H35ClO9 | 50 MG

Supplier: Medchemexpress LLC HY10450A50MG
Dapagliflozin ((2S)-1,2-propanediol, hydrate) is the propanediol hydrate form of the SGLT2 inhibitor dapagliflozin supplied for research use. It is provided as a white to off-white solid with analytically verified purity, suitable for in vitro pharmacology, assay development, and chemical research.
- High purity (99.99% by HPLC)
- White to off-white solid form
- Molecular weight 502.98 g·mol⁻¹
- Chemical formula C24H35ClO9
- Intended for research use only
- Available in small research pack sizes (e.g., 50 mg)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.