Learn More
Medchemexpress LLC Dapagliflozin impurity 112 | 872980-38-6 | 562.99 | C28H31ClO10 | 10 MG

Supplier: Medchemexpress LLC HYI111810MG
Dapagliflozin impurity 112 is a characterized impurity of the SGLT2 inhibitor dapagliflozin, supplied as a reference standard for research and analytical use. It is intended for use as a drug intermediate or analytical standard and is labeled for research use only.
- used as a drug intermediate or analytical standard.
- intended for research use only.
- cas number 872980-38-6.
- molecular formula C28H31ClO10.
- molecular weight 562.99 g/mol.
- storage (powder): -20°C for 3 years, 4°C for 2 years.
- storage (in solvent): -80°C for 6 months, -20°C for 1 month.
- commonly supplied as a 10 mg quantity.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.