Learn More
Medchemexpress LLC Dapagliflozin propanediol hydrate | 960404-48-2 | MFCD28167768 | ≥98.0% | 503.0 g·mol⁻1 | C24H35ClO9 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY10450A10MM 1ML
Encompass_Preferred
Dapagliflozin propanediol hydrate is the propanediol monohydrate form of the SGLT2 inhibitor dapagliflozin, provided for research use in cellular and biochemical studies. It is commonly supplied as a concentrated stock solution (example: 10 mM) for in vitro assays, and the hydrate salt aids handling and stability compared with the free compound.
  • SGLT2 inhibition mechanism useful for glucose transport and metabolic disease research.
  • Ready-to-use stock solution format simplifies preparation for cell-based assays.
  • Hydrate salt form enhances stability and solubility in polar solvents.
  • High analytical purity suitable for biochemical and pharmacology experiments (≥98.0% HPLC).
  • Compatible with common in vitro assay workflows and low-temperature storage.
Catalog No. 50-002-41800