Learn More
Cayman Chemical DaphnoretIn 10mg
SDP
Supplier:  Cayman Chemical 2640810 MG
Encompass_Preferred
A coumarin with diverse biological activities; induces aggregation of washed rabbit platelets (EC50 = 17.2 μM); active against RSV in a plaque reduction assay using HEp-2 cells (IC50 = 5.87 μg/ml); halts the cell cycle at the G2/M phase, induces apoptosis, and inhibits proliferation of HOS cells (IC50 = 3.89 μM); reduces proliferation, invasion, and migration of HCT116 colon cancer cells
Catalog No. 50-293-3543