Learn More
TARGETMOL CHEMICALS INC Daphnoretin 25MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T4S133525MG
Encompass_Preferred
Also available in 1 mL, 5 mg, 10 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. 1. Daphnoretin (Thymelol) has strong antiviral activity, mainly on the later phase of the replication cycle. 2. Daphnoretin has anticancer effects on leukemia, osteosarcoma and uterine cervix cancer cells, might through promoting apoptosis in a mitochondria-mediated way. 3. Daphnoretin causes death of HOS cells by blocking cells successively in G2/M phases and activating the caspase-3 pathway. Purity 99.8%
Catalog No. 50-287-6769