Learn More
Cayman Chemical DAPK SubstrAT PeptIdetrIfl 1mg
SDP
Supplier:  Cayman Chemical 352211 MG
Encompass_Preferred
A DAPK1 peptide substrate; has been used as a target substrate for the quantification of DAPK1 activity
Catalog No. 50-291-1118