Learn More
Medchemexpress LLC DAPK3/ZIPK Human s 50ug | 50ug
SDP
Supplier:  Medchemexpress LLC HYP7728650UG
Encompass_Preferred
The DAPK3/ZIPK protein is a multifunctional serine/threonine kinase that complexly regulates apoptosis autophagy transcription translation and actin cytoskeletal reorganization Its key roles include coordinating apoptotic and autophagy signaling influencing smooth muscle contraction and regulating starvation-induced autophagy DAPK3/ZIPK Protein Human (sf9 GST) is the recombinant human-derived DAPK3/ZIPK protein expressed by Sf9 insect cells with N-GST labeled tag
Catalog No. 50-001-86589