Learn More
Medchemexpress LLC HY-B0688 500mg Medchemexpress, Dapsone CAS:80-08-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB0688500MG
Encompass_Preferred
Medchemexpress, HY-B0688 500mg Dapsone CAS:80-08-0 Dapsone (4,4′-Diaminodiphenyl sulfone) is an orally active and blood-brain penetrant sulfonamide antibiotic with bacteriostatic, antimycobacterial and antiprotozoal activities. Dapsone exerts effective antileprosy activity and inhibits folate synthesis in cell extracts of M. leprae. Dapsone is used for dermatologic disorder research, including leprosy, dermatitis herpetiformis, acne vulgaris et al. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2642