Learn More
TARGETMOL CHEMICALS INC DAPT 25MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T620225MG
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. DAPT (LY-374973) is a novel (gamma)-secretase inhibitor which reduces the A(beta) production (IC50 20 nM in HEK 293 cells) and A(beta)42 levels (IC50 200 nM). purity: 99%
Catalog No. 50-257-9490