Learn More
Selleck Chemical LLC DAPT (GSI-IX) 10mM/1mL 208255-80-5 LY-374973
SDP
Supplier:  Selleck Chemical LLC S221510MM/1ML
Encompass_Preferred
DAPT (GSI-IX, LY-374973) is a novel ?-secretase inhibitor, which inhibits A? production with IC50 of 20 nM in HEK 293 cells. DAPT enhances the apoptosis of human tongue carcinoma cells and regulates autophagy. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-2174