Learn More
Medchemexpress LLC Daptomycin | 103060-53-3 | >99.7% | C72H101N17O26 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYB010850MG
Encompass_Preferred
Daptomycin is a lipopeptide antibiotic characterized by its rapid in vitro bactericidal activity specifically against gram-positive organisms. This compound plays a crucial role in anti-infection pathways, functioning as an effective bacterial inhibitor for research applications.
  • Lipopeptide antibiotic
  • Rapid in vitro bactericidal activity
  • Effective against gram-positive organisms
  • Inhibitor target
  • Suitable for research use
Catalog No. 50-004-32442