Learn More
AdipoGen DAR-4
SDP
Supplier:  AdipoGen CDXD0276M001
Encompass_Preferred
Chemical. CAS 339527-78-5. Formula C24H24N4O3. MW 416.48. Synthetic Photo-stable fluorescent probe for the detection of nitric oxide NO.
Catalog No. 50-256-1147