Learn More
TARGETMOL CHEMICALS INC DARAPLADIB 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T610910MG
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Darapladib (SB-480848)(IC50=0.25 nM) is a substituted pyrimidone with inhibitory activity towards lipoprotein-associated phospholipase-A2 (Lp-PLA2). purity: 99%
Catalog No. 50-257-9552