Learn More
Medchemexpress LLC Daratumumab PBS 10mg | 10mg
SDP
Supplier:  Medchemexpress LLC HYP9915A10MG
Encompass_Preferred
Daratumumab (PBS) (Anti-Human CD38) is the first-in-class human-specific anti-CD38 monoclonal antibody (IgG1) Daratumumab (PBS) has anti-multiple myeloma (MM) effect Daratumumab (PBS) impairs MM cell adhesion which results in an increased sensitivity of MM to proteasome inhibition[1][2]
Catalog No. 50-003-22838