Learn More
Eurofins Discoverx PathHunter Darbepoetin Alfa Bioassay Kit, 10pl. (93-0965Y3-00020)
Supplier:  Eurofins Discoverx 930965Y300020
Encompass
The PathHunter Darbepoetin Alfa Bioassay kit provides an easy-to-use cell based assay to measure drug potency and detect neutralizing antibodies. This bioassay assesses erythropoetin or darbepoetin alfa-induced dimerization of EPOR receptors. This product uses EFC technology. The cell line included overexpresses EPOR target protein fused to PK alongside EPOR target protein fused to EA. Upon ligand-induced dimerization of EPOR, detectable EFC signal is produced. Bioassay kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, control agonist, detection reagent, and assay plates. For detailed product information, User manual and COA, visit www.discoverx.com/bioassay.
Catalog No. 50-200-1699
Only null left
Add to Cart
Request a Quote