Learn More
Cayman Chemical DarglItazon 10mg
SDP
Supplier:  Cayman Chemical 1768110 MG
Encompass_Preferred
A TZD that increases the sensitivity of cells to insulin, stimulating insulin-induced PI3K activity at submicromolar concentrations; increases insulin effectiveness in obese NIDDM subjects; promotes adipocyte differentiation by up-regulating the expression of UCP-2
Catalog No. 50-296-5478