Learn More
Medchemexpress LLC Darifenacin | 133099-04-4 | 99.8% | 426.55 | C28H30N2O2 | 10 MG

Supplier: Medchemexpress LLC HYA003310MG
Darifenacin is a selective M3 muscarinic receptor antagonist supplied as a research-grade small molecule for in vitro and in vivo studies investigating muscarinic receptor pharmacology and related biological effects.
- Selective M3 muscarinic receptor antagonist
- High purity (99.8%)
- Molecular weight 426.55
- Chemical formula C28H30N2O2
- Packaged as 10 mg for research use
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.