Learn More
Selleck Chemical LLC Darifenacin E6020-100MG
SDP
Supplier:  Selleck Chemical LLC E6020100MG
Encompass_Preferred
Darifenacin [( )-UK-88525] is a potent specific and competitive M3 selective receptor antagonist (M3 SRA) that has been shown to have high affinity and selectivity for the M3 receptor It significantly improves major overactive bladder (OAB) symptoms with proven efficacy tolerability and safety
Catalog No. 50-000-43052