Learn More
Cayman Chemical DarIfenacIn-d4 5mg
SDP
Supplier:  Cayman Chemical 337015 MG
Encompass_Preferred
An internal standard for the quantification of darifenacin by GC- or LC-MS
Catalog No. 50-294-7593