Learn More
Apexbio Technology LLC Darifenacin HBr, 10mg. Cas: 133099-07-7 MFCD: MFCD08141803. Selective M3 muscarinic receptor antagonist
SDP
Supplier:  Apexbio Technology LLC B160010
Encompass_Preferred
DarifenacinHBr(UK88525) is a selective M3 muscarinic receptor antagonist with pKi of 8.9 [1]. Themuscarinic acetylcholine receptor, also known as cholinergic/acetylcholine receptor M3, or themuscarinic 3, is amuscarinic acetylcholine receptorencoded by the human geneCHRM3. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5271