Learn More
Selleck Chemical LLC Darifenacin HBr S3144-1g
SDP
Supplier:  Selleck Chemical LLC S31441G
Encompass_Preferred
Darifenacin HBr (UK-88525) is a selective M3 muscarinic receptor antagonist with pKi of 8 9
Catalog No. 50-311-0731