Learn More
Medchemexpress LLC Darifenacin hydrobromide | 133099-07-7 | MFCD08141803 | 99.8% | C28H31BrN2O2 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYA001250MG
Encompass_Preferred
Darifenacin hydrobromide is a selective and orally active M3 muscarinic receptor (M3R) antagonist. It exhibits high specificity for M3R, binding >20-fold more specifically than to other muscarinic receptors. This compound is valuable for studying urinary incontinence and other symptoms associated with an overactive bladder. Additionally, it has demonstrated potential in inhibiting tumor growth in colorectal cancer cells, suggesting anti-tumor effects.
  • Selective and orally active M3 muscarinic receptor antagonist
  • High specificity for M3R
  • Useful in studying urinary incontinence and overactive bladder
  • Inhibits tumor growth in colorectal cancer cells
  • Exhibits anti-tumor effects
Catalog No. 50-004-33302