Learn More
Medchemexpress LLC Darinaparsin | 69819-86-9 | ≥98.0% | 411.31 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY149435MG
Encompass_Preferred
Darinaparsin (ZIO-101), an organic arsenical, is a mitochondrial-targeted agent that induces apoptosis in cancer cells and exhibits anticancer effects.
  • Cytotoxic to prostate cancer cell lines and fresh prostate cancer cells at low micromolar concentrations.
  • Inhibits tumor-initiating cells (TIC) subpopulations.
  • Inhibits growth of the castrate-resistant Du145 prostate tumor propagated as a xenograft in mice.
  • Inhibits the tumor-initiating potential of prostate cancer cells.
Catalog No. 50-003-60248