Learn More
Medchemexpress LLC Darinaparsin (S-dimethylarsino-glutathione) | 69819-86-9 | MFCD17215931 | ≥98.0% | 411.31 g/mol | C12H22AsN3O6S | 10 MG

Supplier: Medchemexpress LLC HY1494310MG
Darinaparsin is an organic arsenical research compound (S-dimethylarsino-glutathione) with mitochondrial-targeted activity that induces apoptosis in cancer cells. It is used in preclinical and translational studies to evaluate anticancer mechanisms and to explore combination therapies.
- Mitochondrial-targeted mechanism of action.
- Induces apoptosis in multiple cancer cell types.
- Suitable for preclinical and translational anticancer research.
- High purity appropriate for research applications.
- Molecular formula C12H22AsN3O6S and molecular weight 411.31 g/mol.
- CAS number 69819-86-9 for unambiguous identification.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.